• USA Home
  • Z381500 - Heating blocks for Techne® Dri-Block® heater
EMAIL THIS PAGE TO A FRIEND
Z381500 Sigma
Heating blocks for Techne® Dri-Block® heater
Plastic spacer block
Purchase
Description
General description
Interchangeable blocks machined of aluminum for rapid heat transfer. Close tolerances assure efficient, rapid heating. Each block has a threaded thermometer hole for independent temperature monitoring, which also fits the block extraction tool.
Two spacer blocks will center a single heating block in a two-block heater
Legal Information
Dri-Block is a registered trademark of Techne, Inc.
Techne is a registered trademark of Techne, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
Z381403 Techne® Dri-Block® heater, Model DB-3D, AC/DC input 240 V AC
Z381373 Techne® Dri-Block® heater, Model DB-3, AC/DC input 240 V AC
Z381330 Techne® Dri-Block® heater, Model DB-2A, AC/DC input 240 V AC
Z381322 Techne® Dri-Block® heater, Model DB-2A, AC/DC input 120 V
Z381365 Techne® Dri-Block® heater, Model DB-3, AC/DC input 120 V
Z381381 Techne® Dri-Block® heater, Model DB-3D, AC/DC input 120 V
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?