• USA Home
  • Z381837 - Ultramicro pipette tips
EMAIL THIS PAGE TO A FRIEND
Z381837 Sigma
Ultramicro pipette tips
capacity 10 μL, non-sterile, Microcapillary, flat tip
Purchase
Properties
Related Categories Automated Sequencing, Equipment and Accessories, Labware, Liquid Handling Equipment, Miscellaneous Pipette Tips,
material   polypropylene
sterility   non-sterile
packaging   tray of 10 × 20 tips
capacity   10 μL
Description
Application
For loading sample wells in 0.2 mm sequencing gels.
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?