• USA Home
  • Z381993 - Whatman® Nytran SuPerCharge (SPC) TurboBlotter refills - large
EMAIL THIS PAGE TO A FRIEND
Z381993 Sigma
Whatman® Nytran SuPerCharge (SPC) TurboBlotter refills - large
Nytran SPC 0.45 μm TurboBlotter refill, 20 x 25 cm, 1/pk
Synonym: blotting membrane, nylon membrane
Purchase
Properties
Related Categories Blotters, Blotting Membranes and Paper, Electrophoresis Supplies, Labware, TurboBlotter Rapid Downward Transfer System and Nytran refills,
packaging   pack of 1
mfr. no.   Whatman 10416326
  Whatman Article No., 28415395 (US reference)
size   20 cm × 25 cm
suitability   suitable for (high-resolution transfer of DNA and RNA; nucleic acid transfer)
Description
Compatibility
TurboBlotter Refills, Large
Nytran SPC, wicks and blotter stacks for five transfers from indicated gel size.
Note: Replacement packet of Turboblotter consumables, sufficient for 5 transfers.
Other Notes
Global SIAL customers (excluding Japan) can now order any product from the GE Healthcare Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
This product has been replaced by WHA10416326-SIGMA | Whatman Nytran SuPerCharge (SPC) TurboBlotter refills - large, Nytran SPC 0.45 μm TurboBlotter refill, 20 x 25 cm, 1/pk
Legal Information
Nytran is a trademark of GE Healthcare
Whatman is a registered trademark of GE Healthcare
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
WHA10416326 Whatman® Nytran SuPerCharge (SPC) TurboBlotter refills - large, Nytran SPC 0.45 μm TurboBlotter refill, 20 x 25 cm, 1/pk
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?