• USA Home
  • Z382205 - Corning® 384 well plates
EMAIL THIS PAGE TO A FRIEND
Z382205 Sigma
Corning® 384 well plates
black (tissue-culture treated), sterile
Purchase
Properties
material   polystyrene
  black (tissue-culture treated)
description   For fluorescence assays
sterility   sterile
feature   lid
mfr. no.   Corning 3709
size   384 wells
Description
General description
These 384 well plates are compatible with leading readers and integrate into high-throughput applications.
Legal Information
Corning is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?