• USA Home
  • Z382477 - Multiwell Plates, Streptavidin coated

EMAIL THIS PAGE TO A FRIEND
Z382477 Sigma

Multiwell Plates, Streptavidin coated

Purchase

Properties

material   polystyrene
color   white
capacity   20-25 pmol/well (Biotin)
storage temp.   2-8°C

Description

Application

Use for biotin-linked screening studies.

Features and Benefits

These 96-well, flat-bottom polystyrene plates are coated on the bottom and wall up to a working volume of 100 μl (±5% well-to-well).
Bioburden (bacterial and fungal) and endotoxin tested
Blocked to prevent non-specific binding up to a volume of 100 μl
Available in three formats:
  • clear, for colorimetric assays
  • white clear bottom for chemiluminescent assays
  • black clear bottom for fluorescent assays

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?