| Related Categories | Absorption Cells, Analytical Reagents, Analytical/Chromatography, Cuvettes, Flow-through Cells, |
| material | Suprasil® quartz |
| mfr. no. | Hellma 178-710-QS |
| limit | 200-2,500 nm spectral range |
| H × W × D | 35 mm × 12.5 mm × 12.5 mm |
| aperture center H | 8.5 mm , aperture |
| chamber volume | 80 μL |
| pathlength | 10 mm |
2 windows polished aperture.
flowthrough cells with 2 screw connectors M 6 X 1 and FEP tubes, aperture 3mm diameter
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?