• USA Home
  • Z604534 - BRAND® multiwell PCR plate, 96 well
EMAIL THIS PAGE TO A FRIEND
Z604534 Sigma
BRAND® multiwell PCR plate, 96 well
Purchase
Properties
Related Categories 96 Well Plates, Labware, Multiwell Plates, Other 96-Well Multiwell Plates
availability   not available in USA and Canada
Description
General description
PP, non-skirted, PCR plates consisting of 96 PCR tubes (0.2 mL). Ultra-thin wall design facilitates rapid heat transfer. The smooth vessel interior minimizes the binding of enzymes and nucleic acids to the walls. Raised rims of wells in the standard profile provide tight seal with self-adhesive sealing films.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
BR781400 BRAND® 96-well PCR plate, semi-skirted, clear, cut corner A1, standard profile
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?