Z604747 Sigma
Polaroid® GelCam® camera system
Electrophoresis Filter Kit
DOWNLOAD MSDS (PDF)
Description
General description
The Polaroid GelCam Camera system is used to make instant photographic records from light-emitting sources such as light boxes, video screens or CRT screens.
Features include:
• Hand-held operation
• No focusing required
• Four element f4.5 glass lens
• Shutter speeds: 1/125 sec to 1 sec and Bulb
• Easily interchangeable hoods
• Pistol grip single-stroke shutter release
• Rugged construction
• Snap-on hoods of various sizes determine the photographic area
Legal Information
GelCam is a registered trademark of Polaroid Corp.
Polaroid is a registered trademark of Polaroid Corp.
Price and Availability
Customers Also Viewed
Electrophoresis hood with filter holder, diag. 7 in.
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?