• USA Home
  • Z605603 - Eppendorf® PCR film, self adhesive
EMAIL THIS PAGE TO A FRIEND
Z605603 Sigma
Eppendorf® PCR film, self adhesive
Purchase
Properties
Related Categories Eppendorf, Eppendorf PCR, Eppendorf PCR Caps & Seals, Labware, Manufacturer Browser,
mfr. no.   Eppendorf®, 0030 127.811 (European)
  Eppendorf®, 951023019 (USA)
availability   not available in USA and Canada
Description
General description
• Effective adhesive, may be removed from plate without residue.
• Transparent film for easy visual monitoring of samples.
• Human genomic DNA, DNase, RNase and PCR Inhibitor free.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?