• USA Home
  • Z605670 - Eppendorf® exchangeable thermoblocks, with rack and coolpack
EMAIL THIS PAGE TO A FRIEND
Z605670 Sigma
Eppendorf® exchangeable thermoblocks, with rack and coolpack
accepts 24 microtubes 2 mL
Purchase
Description
General description
Exchangeable Thermoblocks for use with the Thermomixer R (T3317) and ThermoStat Plus. With 24 position work rack and 0°C cooling pack-designed to keep samples at 0°C on the lab bench for approximately three hours.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Articles
EZ-Fit™ Filtration Products for Bioburden Testing
The EZ family of products has been designed to optimize and streamline the bioburden workflow. The family includes:
Keywords: Filtration
HybriScan™ Rapid Test Systems: Rapid detection, identification and quantification of microorganisms in beverages, food and water
The new HybriScan Test System, which uses sandwich hybridization, provides fast, sensitive and reliable detection, identification and quantification of spoilage and pathogenic microorganisms in bever...
Jvo Siegrist
AnalytiX Volume 8 Article 3
Keywords: Enzyme-linked immunosorbent assay, Microbiology, Nucleic acid hybridization, Polymerase chain reaction, Purification, Transcription
Microbiology Introduction
An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins
Related Content
HybriScan Rapid Test Systems for Microorganisms in Food & Beverages
- Introduction - Benefits over conventional detection methods and PCR - Table with advantages over other detection techniques - Principles of the HybriScan method - Sensitivity, specificity, flexibil...
Keywords: Cell disruption, Clinical, Detection methods, Enzyme-linked immunosorbent assay, Food Safety, Nucleic acid hybridization, Polymerase chain reaction, Purification, Transcription
HybriScan Rapid Test Systems
Das neue HybriScan Testsystem, welches auf der Sandwich-Hybridisierung basiert, bietet einen schnellen, empfindlichen und zuverlässigen Weg Verderbsmikroorganismen und pathogenen Keime in Getränke, N...
Keywords: Cell disruption, Enzyme-linked immunosorbent assay, Nucleic acid hybridization, Polymerase chain reaction, Transcription
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?