• USA Home
  • Z605751 - Eppendorf® Research® pipette, fixed volume
EMAIL THIS PAGE TO A FRIEND
Z605751 Sigma
Eppendorf® Research® pipette, fixed volume
100 μL volume
Purchase
Properties
Related Categories Eppendorf, Eppendorf Pipettors, Eppendorf Pipettors & Tips, Labware, Liquid Handling Equipment,
material   yellow control button
description   use 200 μl pipette tips
feature   electronic: no
  large operating button for fill and blow-out with separate tip ejection.
  lower part autoclavable
  single-channel
mfr. no.   Eppendorf®, 022472003 (USA)
  Eppendorf®, 3112 000.053 (European)
  Eppendorf®, 3112 000.053
availability   not available in USA and Canada
measuring range   100 μL volume
volume   100 μL
Description
Legal Information
Eppendorf Research is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?