• USA Home
  • Z606219 - Eppendorf® exchangeable thermoblocks, with rack and coolpack
EMAIL THIS PAGE TO A FRIEND
Z606219 Sigma
Eppendorf® exchangeable thermoblocks, with rack and coolpack
thermoblock for 24 x 0.5 mL tubes, 1/cs
Synonym: PCR tube rack, cool block, cooling block, heat block, heating block
Purchase
Properties
Related Categories Block Heaters, Heating, Labware, Thermomixer blocks for block heaters
packaging   case of 1
  pack of 1
mfr. no.   Eppendorf®, 022670506 (USA)
  Eppendorf®, 5361 000.015 (European)
Description
General description
Exchangeable Thermoblocks for use with the Thermomixer R (T3317) and ThermoStat Plus. With 24 position work rack and 0°C cooling pack-designed to keep samples at 0°C on the lab bench for approximately three hours.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?