• USA Home
  • Z614319 - Vivaspin 2 centrifugal concentrators
EMAIL THIS PAGE TO A FRIEND
Z614319 Sigma
Vivaspin 2 centrifugal concentrators
MWCO 10,000
Purchase
Properties
Related Categories Centrifugal Concentration, Filtration Supplies, Labware, Manufacturer Browser, Sartorius Vivascience,
material   Hydrosart® hydrophilic cellulose
  cellulose triacetate membrane
  polycarbonate housing
sterility   non-sterile
packaging   package of 25 ea
mfr. no.   Sartorius
max. centrifugal force   8000 × g
dead stop volume   8 μL
hold-up volume   10 μL
membrane area   1.2 cm2
tube size   ~15 mL
working volume   2 mL
pore size   10,000 MWCO
Description
General description
The Vivaspin 2 combines the speed of the classic Vivaspin products with low internal surface and membrane area for superior recoveries from very dilute solutions. Unique to the Vivaspin 2 is the choice of directly pipetting the concentrate from the dead stop pocket built into the bottom of the concentrator, or alternatively reverse spinning into the concentrate recovery cap which can then be sealed for storage. Both methods result in near total concentrate recoveries.
• Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
• Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
• Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Legal Information
Hydrosart is a registered trademark of Sartorius AG
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?