• USA Home
  • Z614696 - Vivaspin 20 centrifugal concentrator
EMAIL THIS PAGE TO A FRIEND
Z614696 Sigma
Vivaspin 20 centrifugal concentrator
pore size 0.2 μm
Purchase
Properties
Related Categories Centrifugal Concentration, Filtration Supplies, Labware, Manufacturer Browser, Sartorius Vivascience,
material   polycarbonate housing
  polyethersulfone membrane
description   PES membrane
sterility   non-sterile
packaging   package of 12 ea
mfr. no.   Sartorius
max. centrifugal force   8,000 × g
max. pressure   5 bar
dead stop volume   50 μL
hold-up volume   20 μL
membrane area   6 cm2
tube size   ~50 mL
working volume   20 mL
pore size   0.2 μm
Description
General description
The Vivaspin 20 mL centrifugal concentrator has been developed to offer increased volume flexibility and performance. Vivaspin 20 can be used in either swing bucket of fixed angle rotors (carriers accepting 15 mL and 50 mL bottles). Vivaspin features twin vertical membranes for unparalleled filtration speed and 100X plus concentrations.
Remaining volume is easy to read off the printed scales on the side of the concentrator and the modified dead stop pocket further simplifies direct pipette recover of the final concentrate.
• Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs
• Highest flow rates and recoveries are achieved due to their particular vertical membrane design
• Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin
Note: One step dealing of concentrated sample using diafiltration cup. Gas pressure filtration; for faster processing gas pressure can be combined with centrifugal force.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?