• USA Home
  • Z614742 - Vivaspin 20 pressure head
EMAIL THIS PAGE TO A FRIEND
Z614742 Sigma
Vivaspin 20 pressure head
pkg of 1 ea
Purchase
Properties
Related Categories Centrifugal Concentration, Filtration Supplies, Labware, Manufacturer Browser, Sartorius Vivascience,
material   polycarbonate housing
  polyethersulfone membrane
sterility   non-sterile
packaging   pkg of 1 ea
mfr. no.   Sartorius
tube size   ~50 mL
working volume   20 mL
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?