• USA Home
  • Z615404 - VivaFlow 200 flipflow filtration
EMAIL THIS PAGE TO A FRIEND
Z615404 Sigma
VivaFlow 200 flipflow filtration
pore size 0.2 μm
Purchase
Properties
Related Categories Centrifugal Concentration, Filtration Supplies, Labware, Manufacturer Browser, Sartorius Vivascience,
material   polyethersulfone membrane
mfr. no.   Sartorius
availability   not available in ROW
  not available in USA and Canada
min recirculation volume   <20 mL
hold-up volume   5.3 mL
membrane area   200 cm2
pore size   0.2 μm
Description
General description
Concentrate 250 mL to under 20 mL in just a few minutes or concentrate one liter 50 times in less than 30 minutes. Alternatively use two Vivaflow 200 in parallel and concentrate 5 liters in under 75 minutes. Near total sample recoveries can be expected with most solutions. Desalting or buffer exchange is just as quick when using the optional recirculation assembly available from Sartorius. The economical standard package comes complete with tubing, pressure control gauge, flow restrictor and high pressure pump tubing. All you need is a peristaltic pump capable of handling 6.4 mm O.D. tubing. Should your pump head require larger tubing, link your own peristaltic tube up to the standard product, using the interconnector provided. Vivaflow 200 is easy to clean and can be used repeatedly.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?