• USA Home
  • Z629251 - Vivacell 250 ultrafiltration concentrator membrane inserts
EMAIL THIS PAGE TO A FRIEND
Z629251 Sigma
Vivacell 250 ultrafiltration concentrator membrane inserts
MWCO 50,000
Purchase
Properties
Related Categories Centrifugal Concentration, Filtration Supplies, Labware, Manufacturer Browser, Sartorius Vivascience,
material   polycarbonate housing
  polycarbonate
  polyethersulfone membrane
sterility   non-sterile
packaging   package of 5 ea
mfr. no.   Sartorius
availability   not available in ROW
  not available in USA and Canada
limit   4 bar max. pressure (60 psi)
W × H   116 mm × 235 mm
dead stop volume   600 μL
membrane area   40 cm2
working volume   250 mL
pore size   50,000 MWCO
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?