• USA Home
  • Z640735 - Uniphor electrophoresis units

EMAIL THIS PAGE TO A FRIEND
Z640735 Sigma

Uniphor electrophoresis units

LS-280/85/2, universal horizontal unit with 3 SCE electrodes (1 cathode, 2 anodes)

Purchase

Properties

Related Categories Brenzel, Brenzel Uniphor Electrophoresis Units, Electrophoresis Supplies, Horizontal Gel Electrophoresis Supplies, Labware,
mfr. no.   LS-280/85/2
electrode distance   ≤2 mm × 85 mm
gel size   ≤165 mm (280 mm)
weight   (3.5 kg)
output   ~3000 V, ≤25 mA

Description

General description

UniPhor units can be used for:
Analytical Protein and Peptide Characterization: Denaturing and non denaturing SDS-PAGE gels IEF in ultrathin PAGE gels (Z640727, Z640719 & Z640751)
Preparative Protein and Peptide Separation: Non-denaturating PAGE gels and Agarose gels IEF in Agarose Ampholyte Gels (Z640719 & Z640751)
Molecular Biology: DNA/RNA fragment analysis, SSCP-Electrophoresis, Long Distance-TGGE (needs external cryo-thermostate with temperature ramping functionality), Flat-Bed Sequencing (Z640743 & Z640751).
All units are equipped with SCE (Solid Carbon Electrodes) electrodes, which help avoid any of the known problems of Platinum wire electrodes due to their higher conductivity and larger contact area with the gel.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?