• USA Home
  • Z640794 - UniBlot blotting systems
EMAIL THIS PAGE TO A FRIEND
Z640794 Sigma
UniBlot blotting systems
L/SC-280
Purchase
Properties
Related Categories Blotters, Brenzel, Brenzel Uniblot Blotting Systems, Electrophoresis Supplies, Labware,
mfr. no.   L/SC-280
gel size   ≤280 mm × 280 mm
Description
General description
The UniBlot-unit is a three-piece, horizontal semi-dry blotter used for the electrophoretic transfer of proteins and nucleic acids from polyacrylamide gels to transfer membranes for immobilization of the molecules upon the transfer membrane.
All blotters are equipped with SCE-electrodes, which offer high corrosion resistance and uniquely even distribution of voltage and current.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?