• USA Home
  • Z640824 - Homo Therm plates for electrophoresis units
EMAIL THIS PAGE TO A FRIEND
Z640824 Sigma
Homo Therm plates for electrophoresis units
CK-30.20
Purchase
Description
General description
High performance laminar flow cooling plate made of Ceran-Ceramic compound material with high thermal conductivity (> 150 W/mK), no internal temperature bridging areas, temperature range 4-65 °C, max feed pressure of coolant ca. 1 bar, with 10 mm hose connectors, with positioning grip for gels.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?