• USA Home
  • Z645389 - Universal sample container with spoon cap
EMAIL THIS PAGE TO A FRIEND
Z645389 Sigma
Universal sample container with spoon cap
plain label
Synonym: chemical container, plastic sample, specimen container
Purchase
Properties
Related Categories Bottles and Caps, Containers, Labware, Manufacturer Browser, Plastic Bottles,
mfr. no.   Sterilin 128SC
H   93.1 mm
cap diam.   30.1 mm
volume   30 mL
Description
General description
Disposable plastic (PS) universal sample container with or without labels.
Aseptically manufactured under clean room conditions (class 7 ISO 14644) to exclude microbiological contamination.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?