• USA Home
  • Z645397 - Universal sample container with integral handle
EMAIL THIS PAGE TO A FRIEND
Z645397 Sigma
Universal sample container with integral handle
volume 30 mL
Synonym: sample bottles, specimen container, specimen containers
Purchase
Properties
Related Categories Bottles and Caps, Containers, Labware, Manufacturer Browser, Plastic Bottles,
mfr. no.   Sterilin
H   93.1 mm
cap diam.   30.1 mm
volume   30 mL
Description
General description
Dippas disposable plastic (PS) sample container with integral handle enable sterile sampling and transportation of liquid without the risk of contamination.
Aseptically manufactured under clean room conditions (class 7 ISO 14644) to exclude microbiological contamination.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?