• USA Home
  • Z655643 - Finnpipette® Focus multichannel Plus pipette
EMAIL THIS PAGE TO A FRIEND
Z655643 Sigma
Finnpipette® Focus multichannel Plus pipette
volume 1-10 μL, 12 channel
Purchase
Description
General description
Finnpipette Focus multichannel Pls speeds up repetitive dispensing. The improved design and new mechanical solutions of Finnpette Focus multichannel Plus models improve accuracy and precision. In the low multichannel pipettes, the super blow-out functions ensures accurate delivery or microsized drops.
Redesigned lightweight tip cone module for improved ergonomics. The grippt finger rest lets the pipette balance at the optimum pipetting angle, improving pipettign results. Finnpipette Focus Multichannel Plus is more comfortable and less tiring over periods of repetitive pipettin. The soft-touch tip ejection is more efficient and requires less effort, which is especially important with multichannel pipettes.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?