Z708666 Sigma

Harris micro-punch

complete, I.D. 2 mm, plastic

DOWNLOAD MSDS (PDF)

Synonym: harris micro punch, micro punch

Properties

Related Categories Electrophoresis Supplies, Gel Handling, Harris, Labware, Manufacturer Browser,
material   plastic
mfr. no.   Whatman Article No., 29002391 (US reference)
  Whatman, WB100062
I.D.   2 mm

Description

General description

Multi-purpose sampling tool for standardizing samplings sizes of DNA blood cards to 1.2 mm in diameter. It is a coring tool designed for simultaneous sample extraction, collection and storage in one operation. Rockwell hardened, hand sharpened tips, are designed for long shelf-life and precise repeatable samples with accurate delivery placement.

Price and Availability

Customers Also Viewed

Harris micro-punch

complete, I.D. 1 mm, plastic

Harris micro-punch

complete, I.D. 1.2 mm, plastic

Harris micro-punch

complete, I.D. 3 mm, plastic

Harris micro-punch

replacement tip, I.D. 2 mm

Harris Uni-Core

I.D. 0.5 mm

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?