• USA Home
  • Z720224 - Ma-GEL-in Universal Protein System
EMAIL THIS PAGE TO A FRIEND
Z720224 Sigma
Ma-GEL-in Universal Protein System
complete kit
Purchase
Description
General description
The Ma-GEL-In Universal Protein system. Designed to accommodate numerous gel formats in 8 x 10 cm pre-cast and 10 x 10 cm hand-cast.
Includes the following:
-Buffer Tank
-Gel capture device
-Vented lid
-Power cords
-Buffer dam
-2-place casting system
-Three sets of 10x10 cm glass slides
-Two sets of 0.8 mm spacers/glass holders
-Two, 15-tooth combs
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?