• USA Home
  • Z720402 - Nalgene® centrifuge tubes, Oak Ridge style 3137 with sealing cap
EMAIL THIS PAGE TO A FRIEND
Z720402 Sigma
Nalgene® centrifuge tubes, Oak Ridge style 3137 with sealing cap
nominal capacity 30 mL
Purchase
Properties
Related Categories Centrifugation Supplies, Centrifuge Tubes, Plastic, Labware, Manufacturer Browser, Nalgene,
material   cap (sealing cap)
  polycarbonate
  tube round bottom
  transparent tube
feature   RCF 50,000 × g
packaging   pkg of 10 ea
mfr. no.   Nalgene
nominal capacity   30 mL
Description
General description
Strong and tough like polycarbonate but much more acid- and base-resistant. Can be used up to 50,000 x g in refrigerated and non-refrigerated centrifuges. Must be filled to at least 80% of the total capacity. With sealing cap closure option. Before autoclaving, just set closure on top of the tube without engaging the threads.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Steriflip Filters Units
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?