• USA Home
  • Z721972 - Nalgene® centrifuge tubes, Oak Ridge style 3115
EMAIL THIS PAGE TO A FRIEND
Z721972 Sigma
Nalgene® centrifuge tubes, Oak Ridge style 3115
nominal capacity 10 mL
Purchase
Properties
Related Categories Centrifugation Supplies, Centrifuge Tubes, Plastic, Labware, Manufacturer Browser, Nalgene,
material   polysulfone
  transparent tube
  tube round bottom
feature   RCF 50,000 × g
mfr. no.   Nalgene
nominal capacity   10 mL
Description
General description
Strong and tough like polycarbonate but polysulfone much more acid- and base-resistant. Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Steriflip Filters Units
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?