ISBN-13 978-0415377348
| Related Categories | Books and Software, Labware, Molecular Biology Books More... |
| publication info | M. Dorak, Garland Science, 2006, 333 pp., soft cover |
With a variety of detection chemistries, an increasing number of platforms, multiple choices for analytical methods and the jargon emerging along with these developments, real-time PCR is facing the risk of becoming an intimidating method, especially for beginners. Real-time PCR provides the basics, explains how they are exploited to run a real-time PCR assay, how the assays are run and where these assays are informative in real life. It addresses the most practical aspects of the techniques with the emphasis on ′how to do it in the laboratory′.
1. An Introduction to Real-time PCR
2. Data Analysis and Reporting
3. Relative Quantification
4. Normalisation
5. High-Throughput Primer and Probe Design
6. Quantitative Analysis of Ocular Gene Expression
7. Quantitative Gene Expression by Real-time PCR
8. Real-time PCR Using SYBR Green
9. High-Resolution Melting Analysis for Scanning and Genotyping
10. Quantitative Analyses of DNA Methylation
11. Mitochondrial DNA Analysis
12. Real-time Immuno-PCR
13. Clinical Microbiology
14. Clinical Virology
15. Solid Organ Transplant Monitoring
16. Real-time PCR Applications in Haematology
17. Prenatal Diagnosis in Clinical Genetics for Monogenic Diseases Caused by Single Nucleotide Changes