• USA Home
  • Z739251 - FlipTube microcentrifuge tubes

EMAIL THIS PAGE TO A FRIEND
Z739251 Sigma

FlipTube microcentrifuge tubes

polyopropylene, blued, 500/cs

Synonym: microfuge tubes

Purchase

Properties

Related Categories 1.0 to 2.0 mL microcentrifuge tubes, Centrifugation Supplies, GEMU FlipTubes for PCR Sample Prep, Labware, Microcentrifuge Tubes,
material   cap (lid)
  polypropylene
feature   RNase and DNase free
  autoclavable
  disposable
  fits all standard centrifuges; centrifugation 20,000 x g
packaging   case of 500
mfr. no.   GEMU FlipTube
  GEMU FlipTube
capacity   1.5 mL

Description

General description

FlipTubes can be opened by applying slight pressure to the rear end of the lid.

Benefits: A significantly reduced risk of cross-contamination between the user′s fingers and the interior of the tube.

• Easy one hand operation, even when standing in a rack
• Graduated: 0.1, 0.5, 1 and 1.5 mL
• Large frosted writing surfaces on both top and side for easy identification
• Piercing area in lid
• Made of high quality, autoclavable polypropylene
• Manufactured in clean rooms
• High thermal and chemical resistivity
• Stands centrifugation to 20,000 x g
• Fits all standard centrifuges
• Available in bags of 500 tubes

Dimensions: Volume 1.5 mL x Inner Diameter 8.2 mm x Outer Diameter 10 mm x Height 39.0 mm.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

Metagenomics and Quantitative PCR

The highly variable regions within the 16S ribosomal RNA (rRNA) gene sequences are being exploited with PCR and metagenomic sequencing to characterize the microbial strains within the gastrointestina...
Chloe McClanahan
Biofiles Edition 6.2
Keywords: Gastrointestinal, Nucleic acid hybridization, Polymerase chain reaction, Polymerase chain reaction - quantitative, Probiotics, Sequences, Sequencing, Usage

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?