• USA Home
  • Z740366 - Urisafe® urine collection container
EMAIL THIS PAGE TO A FRIEND
Z740366 Sigma
Urisafe® urine collection container
with HCL preservative, volume 4 L
Purchase
Properties
Related Categories Containers, Labware, Sample Collection More...
material   high-density polyethylene container
  polypropylene cap
packaging   pack of 30 ea
mfr. no.   Simport, B350-4HCL
volume   4 L
Description
Other Notes
This model offers the convenience of being pre-filled with 30 ml 6N HCL. All patient instructions are clearly detailed on label. Each container is individually wrapped.
Legal Information
Urisafe is a registered trademark of Simport
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?