• USA Home
  • Z740425 - HistoTainer I prefilled specimen container

EMAIL THIS PAGE TO A FRIEND
Z740425 Sigma

HistoTainer I prefilled specimen container

tamper evident, volume 20 mL

Purchase

Properties

Related Categories Containers, Labware, Sample Collection More...
material   polyethylene cap
  polypropylene container
description   50% filled with 10% neutral buffered Formalin
packaging   pack of 96 ea
mfr. no.   Simport, M960-20FMA
volume   20 mL

Description

General description

Especially designed for collection, transport and storage of histology specimens, Simport offers shatter resistant polypropylene containers, eliminating most problems of leakage and evaporation. The magenta lids are ribbed for easy opening when hands are wet or gloved while the jars are stackable for easy, safe storage and translucent to allow specimens to be viewed without opening. The closures are manufactured from virgin polyethylene with a unique integrated leak-resistant seal. Container′s vertical walls offer excellent rigidity.

The uniqueness of the HistoTainer I is that it incorporates an innovative tamper evident screw cap ensuring your peace of mind during transport or storage situations where someone might have manipulated the specimen without your prior knowledge. Can also be used without using the tamper evident locking mechanism.

Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used in manufacture are free from latex. All containers are 95 kPa compliant. Available in many sizes from 20 to 120 ml. Packaging in trays of 24, cases of 96.

The Simport HistoTainer I is half filled with 10% Neutral Buffered Formalin as a fixative. 10% Neutral Buffered Formalin penetrates quickly, but fixes slowly. The Simport Formalin is enhanced by a buffering capacity optimizing histological results by light microscopy and immunohistochemistry.

Legal Information

HistoTainer is a trademark of Simport

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?