• USA Home
  • Z740453 - Isohelix DNA Buccal Swabs, ethylene oxide treated
EMAIL THIS PAGE TO A FRIEND
Z740453 Sigma
Isohelix DNA Buccal Swabs, ethylene oxide treated
with 5 mL tube and cap, pkg of 100 ea (individually wrapped)
Purchase
Properties
Related Categories Labware, Sample Collection, Tubes More...
packaging   pkg of 100 ea (individually wrapped)
Description
General description
The Isohelix range of DNA Buccal Swabs has been specifically designed to give increased yields of high quality buccal cell and genomic DNA. Buccal cell sampling offers a viable alternative to blood collection and, together with the Isohelix range of DNA Isolation and stabilization kits, enables high yields of pure, intact DNA to be collected easily and quickly.
Isohelix DNA swabs are suitable for both human and veterinary use and offer significant advantages over other DNA swab designs in terms of efficiency of cell collection by the unique swab matrix, which combined with a quick release surface, maximizes yields of DNA. Swab batches are ethylene oxide treated and routinely tested for human DNA contamination.
Features:
• High yielding swabs for buccal cell DNA collection
• Unique swab matrix greatly improves DNA yields
• Alternative to blood collection
• Suitable for human and veterinary use
• Easy to handle and quick to use
• Available in single and twin sterile packs
Applications:
• Research Genotyping Applications
• Veterinary Genotyping and Diagnostics
• Paternity and Heredity Services
• Forensics and DNA Population Studies
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?