• USA Home
  • Z740461 - Isohelix Dri-Capsules
EMAIL THIS PAGE TO A FRIEND
Z740461 Sigma
Isohelix Dri-Capsules
for long term DNA stability on SK1 swabs, pkg of 50 ea (individually foil wrapped)
Purchase
Properties
Related Categories Labware, Sample Collection, Tubes More...
packaging   pkg of 50 ea (individually foil wrapped)
Description
General description
Isohelix Dri-Capsules
Designed for use with Isohelix SK-1 swabs, these silica gel capsules offer an easy to use alternative to chemical stabilisation for long term stability of the buccal DNA on the swab head prior to isolation.
Features:
• Long term stability at room temperature
• Liquid free alternative to DSK Kit
• Easy to handle by patients and professionals
• Designed for use with SK-1 swab packs
• Ideal for remote sample collection
Long term stability studies show that the stability of DNA samples on SK1 swabs stored with Isohelix Dri-Capsules is maintained over a period of at least 3 years. The ongoing study is expected to confirm an increased period of stabilization well in excess of the 3 years tested so far.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?