• USA Home
  • Z756687 - Tissue disaggregator
EMAIL THIS PAGE TO A FRIEND
Z756687 Sigma
Tissue disaggregator
12 wells, pkg of 1 ea
Purchase
Properties
Related Categories Bel Art Tissue Sample Prep, Bel-Art, Cell Culture Supplies, Labware, Manufacturer Browser,
feature   12 wells
packaging   pkg of 1 ea
mfr. no.   Bel-Art, F37844-0000
overall L × W × H   13.2 cm × 9.0 cm × 2.6 cm
well D × diam.   2.0 cm × 1.8 cm
Description
General description
Achieve effective and economical disaggregation of soft tissue to produce cell isolates for culture or flow cytometry. Unlike single cup designs, twelve wells speed processing time and reduce per sample costs by allowing multiple cell isolates to be produced at one time. Use a syringe plunger or suitable device to press soft tissues through the screen thereby disrupting the organ structure and producing cell isolates.
• Provides increased cell viability compared to alternate methods
• Permanently affixed screens at the bottom of each well won′t trap tissues or allow non-screened tissues to pass into the collection plate
• Constructed of FDA-grade polymethylpentene with 70 mesh stainless steel screens, the disaggregator is completely clear for full visibility of the wells
• Overall Dimensions: 13.2 L x 9.0 W x 2.6 cm H (5.2 x 3.5 x 1")
• Individual Wells: 1.8 diameter x 2.0 cm depth (0.7 x 0.8")
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?