EMAIL THIS PAGE TO A FRIEND
11880-U Supelco

Coated GC Packing

phase 0.8% THEED, matrix 80/100 Carbopack C support, bottle of 15 g

Purchase

Properties

Related Categories Analytical/Chromatography, Coated GC Packings, GC Packings, Gas Chromatography (GC), Packed GC Columns and Components More...
packaging   bottle of 15 g
parameter   60-115 °C temp. range
application(s)   gas chromatography (GC): suitable
matrix   80/100 Carbopack C support
matrix active group   0.8% THEED phase
compatibility   for analyte group ethylene oxide (EtO) residues, glycols

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

General description

Ready-to-use coated packing for those wanting to pack their own columns.

The choice of coating material is based on the stationary phase of the GC. THEED (tetrahydroxyethylenediamine) is generally suitable to reduce tailing and modify selectivity on inert adsorbent medium such as Carbopack C.

Application

Gas chromatographic column packed with Carbopack C modified with THEED was used for the determination of diethylene glycol in wine as it is able to elute glycols as untailed peaks.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?