• USA Home
  • 12425 - Metal Packed GC Column (General Configuration)

EMAIL THIS PAGE TO A FRIEND
12425 Supelco

Metal Packed GC Column (General Configuration)

phase 5% Fluorcol, matrix 60/80 Graphitized Carbon Black support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm

  •  eCl@ss 32110502

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Metal Packed GC Columns (General Configuration), Packed GC Columns, Packed GC Columns and Components More...
material   SP alloy column
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   10.0 ft (3.0 m) × 1/8 in. × 2.1 mm
matrix   60/80 Graphitized Carbon Black support
matrix active group   5% Fluorcol phase
column type   packed GC

Description

General description

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.

Packed column made up of stainless steel are basically employed when higher carrier gas flow rates are employed as they can sustain elevated pressure. Stainless steel columns are usually washed with dilute hydrochloric acid, then with water followed by methanol, acetone, methylene dichloride and n-hexane in order to remove any corrosion products and traces of lubricating agents.

Application

Graphitized carbon black packed in a stainless steel GC column maybe used for the analysis of volatile organic compounds (VOCs) present in the environment.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?