• USA Home
  • 12596 - Metal Packed GC Column (fits Varian®)

EMAIL THIS PAGE TO A FRIEND
12596 Supelco

Metal Packed GC Column (fits Varian®)

phase 3% SP-1500, matrix 80/120 Carbopack B support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Metal Packed GC Columns (fit Varian), Packed GC Columns, Packed GC Columns and Components More...
material   stainless steel column
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   10.0 ft (3.0 m) × 1/8 in. × 2.1 mm
matrix   80/120 Carbopack B support
matrix active group   3% SP-1500 phase
column type   packed GC

Description

General description

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column fits Varian 3000, 4000, 6000, and Vista (FID, inj A to det A).

Packed column made up of stainless steel are basically employed when higher carrier gas flow rates are employed as they can sustain elevated pressure. Stainless steel columns are usually washed with dilute hydrochloric acid, then with water followed by methanol, acetone, methylene dichloride and n-hexane in order to remove any corrosion products and traces of lubricating agents.

Application

Carbopack B SP-1500 packed gas chromatographic column maybe used for the analysis of toluene in order to study the activity and three dimensional distribution of toluene degrading Pseudomonas putida in a multispecies biofilm assessed by quantitative in situ hybridization and scanning confocal laser microscopy.

Legal Information

Varian is a registered trademark of Varian Medical Systems, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?