• USA Home
  • 13617-U - Sepabeads® SP-20SS

EMAIL THIS PAGE TO A FRIEND
13617-U Supelco

Sepabeads® SP-20SS

bottle of 100 g

Purchase

Properties

Related Categories Adsorbent Media (Polymeric), Adsorbents (Resins), Analytical/Chromatography, Diaion/Sepabeads, Diaion/Sepabeads/MCI GEL,
packaging   bottle of 100 g
matrix   styrene-divinylbenzene
particle size   50-100 μm
pore size   ~1.01 mL/g pore volume
  260 Å mean pore size
surface area   ~500 m2/g
density   1.30 g/mL at 25 °C (true wet)(lit.)

Description

Application

Polyaromatic adsorbent resin for separation of hydrophobic compounds: small and medium proteins, peptides, amino acids. Excellent for reversed phase applications.

Sepabeads form a polyaromatic adsorbent resin for separation of hydrophobic compounds including small and medium proteins, peptides and amino acids. Sepabeads are excellent for reversed phase applications.

Other Notes

Swelling in toluene = 30%

Legal Information

Sepabeads is a registered trademark of Mitsubishi Chemical Corp.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?