• USA Home
  • 13769 - Metal Packed GC Column (fits PerkinElmer®)

EMAIL THIS PAGE TO A FRIEND
13769 Supelco

Metal Packed GC Column (fits PerkinElmer®)

phase 10% SP-2100, matrix 100/120 SUPELCOPORT support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Metal Packed GC Columns (fit PerkinElmer), Packed GC Columns, Packed GC Columns and Components More...
material   stainless steel column
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
matrix   100/120 SUPELCOPORT support
matrix active group   10% SP-2100 phase
column type   packed GC

Description

General description

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column fits PerkinElmer 8000, 9000, AutoSystem, XL, and Clarus 500 (Not On-Column Injection).

Packed column made up of stainless steel are basically employed when higher carrier gas flow rates are employed as they can sustain elevated pressure. Stainless steel columns are usually washed with dilute hydrochloric acid, then with water followed by methanol, acetone, methylene dichloride and n-hexane in order to remove any corrosion products and traces of lubricating agents.

Legal Information

PerkinElmer is a registered trademark of PerkinElmer Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?