| Related Categories | Analytical/Chromatography, Gas Chromatography (GC), Metal Packed GC Columns (General Configuration), Packed GC Columns, Packed GC Columns and Components More... |
| material | stainless steel column |
| L × O.D. × I.D. | 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm |
| matrix | 80/100 Molecular Sieve 5A support |
| matrix active group | none phase |
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.