EMAIL THIS PAGE TO A FRIEND
13909-U Supelco

Amberchrom® CG300

Amberchrom® CG300M, 50-100 μm

Purchase

Properties

Related Categories Adsorbent Media (Polymeric), Amberlite & Amberchrom, Analytical/Chromatography, Resins & LC Separation Media, Resins and Media More...
packaging   bottle of 100 mL
matrix   styrene-divinylbenzene
particle size   50-100 μm
pore size   0.7 mL/g pore volume
  300 Å mean pore size
surface area   700 m2/g

Description

Application

Polyaromatic resin for rapid adsorption of hydrophobic compounds: surfactants, medium-sized proteins, polypeptides, large peptides, antibiotics. Useful for many reversed phase applications.

Amberchrom® CG300 may be used in separation medium for TLC, paper chromatography and LPLC.

General description

Amberchrom® CG300 is methacrylate resin. It is the highest adsorbing resin for tumour necrosis factor-α (TNF-α), interleukin (IL-8) and C3a against charcoal as reference.

Legal Information

Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1170C 3 / PGII
WGK Germany 
3
Flash Point(F) 
100.4 °F
Flash Point(C) 
38 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?