• USA Home
  • 18912 - Lipid Standards: Fatty Acid Methyl Ester mixtures

EMAIL THIS PAGE TO A FRIEND
18912 Supelco

Lipid Standards: Fatty Acid Methyl Ester mixtures

C20:1 - C20:5 (w/o C20:3)

Purchase

Properties

Related Categories Biochemicals and Reagents, Kits and Mixtures, Lipid Analytical Standards, Lipids, Other Lipid Related Products More...
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

40 mg ampule contains approx. equal amounts by weight of:
cis-11-Eicosenoic acid methyl ester (C20:1)
cis-11,14-Eicosadienoic acid methyl ester (C20:2), 98%
cis-5,8,11,14--Eicosatetraenoic acid methyl ester (C20:4)
cis-5,8,11,14,17-Eicosapentaenoic Acid Methyl Ester (C20:5)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?