• USA Home
  • 18916 - Lipid Standards: Fatty Acid Methyl Ester mixtures

EMAIL THIS PAGE TO A FRIEND
18916 Supelco

Lipid Standards: Fatty Acid Methyl Ester mixtures

C18:0 - C20:0

Purchase

Properties

Related Categories Biochemicals and Reagents, Kits and Mixtures, Lipid Analytical Standards, Lipids, Other Lipid Related Products More...
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

100 mg ampule contains the following with the weight percentages as indicated.
Stearic acid methyl ester (C18:0) 10%
Oleic acid methyl ester (C18:1,cis-9) 20%
Elaidic acid methyl ester (C18:1,trans-9) 20%
Linoleic acid methyl ester (C18:2,cis-9,12) 20%
Linolelaidic acid methyl ester (C18:2,trans-9,12) 20%
Arachidic acid methyl ester (C20:0) 10%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?