• USA Home
  • 18917 - Fatty Acid Methyl Ester Mix

EMAIL THIS PAGE TO A FRIEND
18917 Supelco

Fatty Acid Methyl Ester Mix

C14 - C22,  wt. % (varied conc.), ampule of 100 mg

Synonym: Lipid Standards: Fatty Acid Methyl Ester mixtures

Purchase

Properties

packaging   ampule of 100 mg
concentration    wt. % (varied conc.)
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

100 mg ampule contains the following with weight percentages as indicated.
Methyl esters of
Myristic Acid (C14:0) 4%
Palmitic Acid (C16:0) 10%
Stearic Acid (C18:0) 6%
Oleic Acid (C18:1,cis-9) 25%
Elaidic Acid (C18:1,trans-9) 10%
Linoleic Acid (C18:2,cis-9,12) 34%
Linolelaidic Acid (C18:2,trans-9,12) 2%
Linolenic Acid (C18:3,cis-9,12,15) 5%
Arachidic Acid (C20:0) 2%
Behenic Acid (C22:0) 2%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?