EMAIL THIS PAGE TO A FRIEND
1891 Supelco

Fatty acid standards

Purchase

Properties

Related Categories Biochemicals and Reagents, Kits and Mixtures, Lipid Analytical Standards, Lipids, Other Lipid Related Products More...
packaging   pkg of 100 mg
shipped in   dry ice
storage temp.   −20°C

Description

General description

Our GLC-10 quantitative mix is useful for determining relative retention times and approximating response factors. The mix is carefully prepared by weight percent.

This mixture is now offered as a certified reference material (CRM). Please see product CRM1891 for details and ordering.

Other Notes

100 mg ampule contains approx. equal amounts by weight of:
Palmitic Acid Methyl Ester (C16:0)
Stearic Acid Methyl Ester (C18:0)
Oleic Acid Methyl Ester (C18:1,cis-9)
Linoleic Acid Methyl Ester (C18:2,cis-9,12)
Linolenic Acid Methyl Ester (C18:3,cis-9,12,15)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

CRM1891 F.A.M.E. Mix GLC-10, certified reference material, pkg of 100 mg (Neat)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?