• USA Home
  • 18920-1AMP - F.A.M.E. Mix, C8-C22 Unsaturates

EMAIL THIS PAGE TO A FRIEND
18920-1AMP Supelco

F.A.M.E. Mix, C8-C22 Unsaturates

analytical standard,  wt. % (varied)

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Applications, Fats (fatty acids, FAMEs, glycerides), Edible Oils, Sterols, Fatty Acid Methyl Ester (FAME) Standards,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   pkg of 100 mg
concentration    wt. % (varied)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution
shipped in   dry ice
storage temp.   −20°C

Description

Application

It was used as standard to obtain retention times and also to calibrate chromatogram, during characterizing the difference in fatty acid composition to measure fatty acid profiles from animal tissues using GC method.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools

Price and Availability

Analyte

Description

Product #

Add to Cart

Methyl arachidate 1.9 wt. % SDS    
Methyl behenate 1.9 wt. % SDS    
Methyl decanoate 3.2 wt. % SDS    
Methyl cis-13-docosenoate 1.9 wt. % SDS    
Methyl dodecanoate 6.4 wt. % SDS    
Methyl cis-11-eicosenoate 1.9 wt. % SDS    
Methyl elaidate 2.6 wt. % SDS    
Methyl heptadecanoate 3.2 wt. % SDS    
Methyl linoleate 13 wt. % SDS    
Methyl linolenate 6.4 wt. % SDS    
Methyl myristate 3.2 wt. % SDS    
Methyl myristoleate 1.9 wt. % SDS    
Methyl octanoate 1.9 wt. % SDS    
Methyl oleate 19.6 wt. % SDS    
Methyl palmitate 13 wt. % SDS    
Methyl palmitoleate 6.4 wt. % SDS    
Methyl pentadecanoate 1.9 wt. % SDS    
Methyl stearate 6.5 wt. % SDS    
Methyl tridecanoate 3.2 wt. % SDS    
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

CRM18920 F.A.M.E. Mix, C8 - C22, certified reference material, ampule of 100 mg (Neat)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?