EMAIL THIS PAGE TO A FRIEND
1895 Supelco

Fatty acid standards

Purchase

Properties

Related Categories Biochemicals and Reagents, Kits and Mixtures, Lipid Analytical Standards, Lipids, Other Lipid Related Products More...
shipped in   dry ice
storage temp.   −20°C

Description

Other Notes

100 mg ampule contains approx. equal amounts by weight of:
Palmitic acid methyl ester (C16:0)
Stearic acid methyl ester (C18:0)
Arachidic acid methyl ester (C20:0)
Behenic acid methyl ester (C22:0)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?