• USA Home
  • 1X4400 - Dowex® 1X4 chloride form

EMAIL THIS PAGE TO A FRIEND
1X4400 Supelco

Dowex® 1X4 chloride form

200-400 mesh

Synonym: Dowex® 1 Strongly Basic Anion Exchanger, Chloride form

Purchase

Properties

composition   Moisture, 55-63%
particle size   200-400 mesh
capacity   1.0 meq/mL total capacity

Description

Legal Information

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

428604 Dowex® 1X4 chloride form, chloride form, 200-400 mesh

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?