• USA Home
  • 1X8200 - Dowex® 1X8 chloride form

EMAIL THIS PAGE TO A FRIEND
1X8200 Supelco

Dowex® 1X8 chloride form

100-200 mesh

Purchase

Properties

limit   66 °C max. temp.
cross-linkage   8%
matrix   styrene-divinylbenzene (gel)
particle size   100-200 mesh
operating pH   0 - 14
capacity   1.2 meq/mL total capacity

Description

Application

Strongly basic (type I) anion exchange resin for fine chemical processing; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

217425 Dowex® 1X8 chloride form, chloride form, 100-200 mesh

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?