• USA Home
  • 20238 - Carbotrap® 100 Dynatherm 890, 850

EMAIL THIS PAGE TO A FRIEND
20238 Supelco

Carbotrap® 100 Dynatherm 890, 850

for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU, and OI Analytical Air Tube Desorber or DMP-16

Purchase

Properties

Related Categories Active Sampling, Air Monitoring, Analytical/Chromatography, Dynatherm Standard Sampling Tubes, Sample Preparation & Purification,
material   glass TD tube (non-fritted)
feature   preconditioned
packaging   pkg of 1 ea
mfr. no.   Supelco Carbotrap® 100
I.D. × O.D.   4 mm × 6 mm
matrix   Carbotrap® B graphitized carbon black
suitability   suitable for D6196 per ASTM
compatibility   for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU, and OI Analytical Air Tube Desorber or DMP-16

Description

Application

C5-C12 compounds in air

Legal Information

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC

Dynatherm is a trademark of CDS Analytical

Price and Availability


All labs need water

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

Thermal Desorption Tube Selection Guide

In addition to the general guidelines presented in the tables below view the technical report titled “A Tool for Selecting an Adsorbent for Thermal Desorption Applications” (1,770Kb pdf - T402025), w...
Keywords: Sample preparations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?